Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
1907-33-1,96-47-9
C4H9LiO
80.05
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]
1907-33-1,96-47-9
C4H9LiO
80.05
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide,lithium 2-methylpropan-2-olate,tert-butoxylithium,lithium tert-butylate,lithium tert-butanolate,tert-butyl alcohol, lithium salt,lithium t-butoxide,2-methyl-2-propanol lithium salt,2-propanol, 2-methyl-, lithium salt,lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]
Linear Formula LiOCH(CH3)3
Formula Weight 80.05
Density 0.8600g/mL
Specific Gravity 0.86
Quantity 100mL
Flash Point -11°C
Chemical Name or Material Lithium tert-butoxide

GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger

EINECSNumber : 217-611-5