Lithium tetraborate, 99.6% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1259230

Quantity 250g

 View more versions of this product

Catalog No. AA1259230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12007-60-2
    Physical Form Powder
    Assay Percent Range 99.6% (Metal basis)
    Molecular Formula B4Li2O7
    Formula Weight 169.12
    MDL Number MFCD00011083
    Chemical Name or Material Lithium tetraborate

    Specifications

    Physical Form Powder
    Formula Weight 169.12
    Quantity 250g
    Color White
    Density 2.4g/cm3 at 20°C
    EINECS Number 234-514-3
    Assay Percent Range 99.6% (Metal basis)
    Molecular Formula B4Li2O7
    Appearance White powder
    MDL Number MFCD00011083
    CAS 12007-60-2
    Health Hazard 1 H302-H315-H319-H335: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Lithium tetraborate

    H302-H315-H319-H335: Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit