Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Tetraborate, Flux Grade, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
Packaging:
Poly Bottle

Chemical Identifiers

12007-60-2
Li2B4O7

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Formula Weight 169.11
Packaging Poly Bottle
Specific Gravity 1L = 2.40kg
Quantity 500g
Chemical Name or Material Lithium Tetraborate, Flux Grade
Grade BAKER INSTRA-ANALYZED™ Reagent