Lithium Tetraborate, Flux Grade, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Manufacturer: J.T.Baker 450305

Packaging Poly Bottle
Quantity 2kg

 View more versions of this product

Catalog No. 02-003-222

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12007-60-2
    Molecular Formula Li2B4O7
    Formula Weight 169.11
    Chemical Name or Material Lithium Tetraborate, Flux Grade
    Grade BAKER INSTRA-ANALYZED™ Reagent

    Specifications

    Formula Weight 169.11
    CAS 12007-60-2
    Quantity 2kg
    Packaging Poly Bottle
    Specific Gravity 1L = 2.40kg
    Molecular Formula Li2B4O7
    Chemical Name or Material Lithium Tetraborate, Flux Grade
    Grade BAKER INSTRA-ANALYZED™ Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit