Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Lithium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis)
Click to view available options
Quantity:
0.25g
1g
5g
Specifications
Formula Weight | 345.72 (Anhydrous) |
Physical Form | Crystalline To Liquidous |
Percent Purity | 99.99% |
Grade | Premion™ |
Quantity | 5g |
Chemical Name or Material | Lithium tetrachloroaurate(III) hydrate |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures