Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tetrachlorocuprate, 0.1M in THF, Thermo Scientific Chemicals

Catalog Number p-7049737
Click to view available options
Quantity:
100 mL
500 mL
15489-27-7
Cl4CuLi2
219.226
MFCD00011081
HCJWWBBBSCXJMS-UHFFFAOYSA-J
11074879
dilithium;tetrachlorocopper(2-)
[Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl
15489-27-7
Cl4CuLi2
219.226
MFCD00011081
HCJWWBBBSCXJMS-UHFFFAOYSA-J
11074879
dilithium;tetrachlorocopper(2-)
[Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Orange
Physical Form Liquid
Quantity 100 mL
UN Number UN2056
Solubility Information Difficult to mix.
Formula Weight 219.24
Concentration or Composition (by Analyte or Components) 0.1M in THF
Sensitivity Moisture sensitive
Density 0.910 g/mL
Chemical Name or Material Lithium tetrachlorocuprate
Hazard Category H225-H302-H319-H335-H336-H351
Hazard Statement GHS H Statement
H225-H318-H351-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit