Learn More
Lithium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Description
Lithium tetrafluoroborate is extensively utilized as an electrolyte in primary and secondary cells. It is used in rechargeable Li ion batteries. The electrolytes are non aqueous solutions of lithium tetrafluoroborate in the organic media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 293°C to 300°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 10 g |
Linear Formula | LiBF4 |
UN Number | UN3260 |
Merck Index | 14,5543 |
Solubility Information | Soluble in water and tetrahydrofuran. Slightly soluble is diethyl ether. |
Formula Weight | 93.75 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332-H314-H318 |
Hazard Statement | GHS H Statement H314-H318-H302-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P362-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
EINECSNumber | 238-178-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.