Alfa Aesar™ Lithium tetrafluoroborate, ultra dry, 99.997% (metals basis)

Manufacturer: Alfa Aesar™ 1453206

Quantity 5g

 View more versions of this product

Catalog No. AA1453206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14283-07-9
    Linear Formula LiBF4
    Formula Weight 93.75
    MDL Number MFCD00011087
    UN Number UN3260
    Merck Index 14,5543
    Chemical Name or Material Lithium tetrafluoroborate, Ultra dry

    Specifications

    Formula Weight 93.75
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Quantity 5g
    Health Hazard 1 Danger
    Physical Form Powder
    MDL Number MFCD00011087
    TSCA TSCA
    Sensitivity Hygroscopic
    Linear Formula LiBF4
    CAS 14283-07-9
    UN Number UN3260
    Merck Index 14,5543
    Color White to off-white
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Melting Point 293° to 300°C (dec.)
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Lithium tetrafluoroborate, Ultra dry
    Assay 99.997%, (metals basis)

    GHS H Statement
    H314-H318-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit