Alfa Aesar™ Lithium tetramethylcyclopentadienide, 97+%

Manufacturer: Alfa Aesar™ H27840-03

Quantity 1g

 View more versions of this product

Catalog No. AAH2784003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 82061-21-0
    Molecular Formula C9H13Li
    Formula Weight 128.14g/mol
    MDL Number MFCD00269824
    UN Number UN3391
    Chemical Name or Material Lithium tetramethylcyclopentadienide

    Specifications

    Melting Point >350°C
    Quantity 1g
    CAS 82061-21-0
    Molecular Formula C9H13Li
    Formula Weight 128.14g/mol
    MDL Number MFCD00269824
    UN Number UN3391
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H250-H252
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities; may catch fire.
    Health Hazard 3 GHS P Statement
    P210-P222-P280-P370+P378a-P422a-P420
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not allow contact with air.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store contents under inert gas.
    Store away from other materials.
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
    Chemical Name or Material Lithium tetramethylcyclopentadienide
    Assay ≥97%

    GHS H Statement
    H250-H252
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities; may catch fire.

    GHS P Statement
    P210-P222-P280-P370+P378a-P422a-P420
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not allow contact with air.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store contents under inert gas.
    Store away from other materials.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit