Learn More
Strem, An Ascensus Company CAS# 75965-35-4. 5g. Lithium tetraphenylborate tris(1,2-dimethoxyethane)adduct, 98%. MFCD00013311
SDP
Supplier:  Strem, An Ascensus Company 9305725G
Encompass
CAS# 75965-35-4. 5g. Lithium tetraphenylborate tris(1,2-dimethoxyethane)adduct, 98%. MFCD00013311. Molecular Weight: 326.18 (596.54). Molecular Formula: LiB(C6H5)4∙3CH3OCH2CH2OCH3. Color/form: white to off-white pwdr. Strem# 93-0572. http://www.strem.com/catalog/v/93-0572/
Catalog No. 50-901-13761