Alfa Aesar™ Lithium titanium oxide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 41650A1

Quantity 1kg

 View more versions of this product

Catalog No. AA41650A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12031-82-2
    Formula Weight 109.78
    MDL Number MFCD00016181
    Chemical Name or Material Lithium titanium oxide

    Specifications

    Formula Weight 109.78
    Mesh Size −50 Mesh
    Physical Form Powder
    CAS 12031-82-2
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00016181
    Melting Point 1520° to 1564°C
    Quantity 1kg
    Chemical Name or Material Lithium titanium oxide
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit