Lithium titanium oxide, Off-white powder, Alfa Aesar™

Manufacturer: Alfa Aesar 1341118

Quantity 50g

 View more versions of this product

Catalog No. AA1341118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12031-82-2
    Physical Form Powder
    Molecular Formula Li2TiO3
    Formula Weight 109.78
    MDL Number MFCD00016181
    Synonym Lithium titanate
    Chemical Name or Material Lithium titanium oxide

    Specifications

    Physical Form Powder
    Formula Weight 109.78
    Quantity 50g
    Color Off-white
    Molecular Formula Li2TiO3
    CAS 12031-82-2
    Odor Odorless
    MDL Number MFCD00016181
    Synonym Lithium titanate
    Chemical Name or Material Lithium titanium oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit