Alfa Aesar™ Lithium tri-sec-butylborohydride, 1.0M in THF

As a selective reducing agent

Manufacturer: Alfa Aesar™ 43845AC

Quantity 25mL

 View more versions of this product

Catalog No. AA43845AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38721-52-7
    Molecular Formula C12H28BLi
    Linear Formula LiB[CH(CH3)CH2CH3]3H
    Formula Weight 190.11
    MDL Number MFCD00011708
    UN Number UN3394
    Beilstein 4007327
    Merck Index 14,8427
    Synonym Lithium tri-sec-butylhydridoborate
    Chemical Name or Material Lithium tri-sec-butylborohydride

    Specifications

    Formula Weight 190.11
    Health Hazard 2 GHS H Statement
    H225-H250-H260-H314-H318
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Linear Formula LiB[CH(CH3)CH2CH3]3H
    CAS 38721-52-7
    Synonym Lithium tri-sec-butylhydridoborate
    Concentration 1.0M in THF
    Color Colorless to yellow
    TSCA TSCA
    Density 0.870g/mL
    Merck Index 14,8427
    Health Hazard 1 Danger
    Quantity 25mL
    Flash Point −17°C (1°F)
    UN Number UN3394
    Beilstein 4007327
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    Physical Form Liquid
    Molecular Formula C12H28BLi
    MDL Number MFCD00011708
    Sensitivity Moisture Sensitive
    Chemical Name or Material Lithium tri-sec-butylborohydride

    GHS H Statement
    H225-H250-H260-H314-H318
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit