Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium tri-sec-butylborohydride, 1.0M in THF

As a selective reducing agent

Manufacturer:  Alfa Aesar™ 43845AP

Quantity 500mL

 View more versions of this product

Catalog No. AA43845AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

38721-52-7
LiB[CH(CH3)CH2CH3]3H
MFCD00011708
4007327
Lithium tri-sec-butylhydridoborate
C12H28BLi
190.11
UN3394
14,8427
Lithium tri-sec-butylborohydride

Specifications

38721-52-7
C12H28BLi
190.11
MFCD00011708
4007327
Lithium tri-sec-butylhydridoborate
Colorless to yellow
−17°C (1°F)
Lithium tri-sec-butylborohydride
Liquid
LiB[CH(CH3)CH2CH3]3H
0.870g/mL
UN3394
14,8427
1.0M in THF
500mL
Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H250-H260-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit