Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tri-sec-butylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
AcroSeal™ Glass Bottle in Tin Can
38721-52-7,109-99-9,1113-78-6
C12H28BLi
MFCD00011708
lithium tri-sec-butylhydroborate,l-selectridesolution,lithium tri-sec-butylborohydride solution,l-selectride™ solution,lithium tri butan-2-yl boron 1-,lithium tri-sec-butylborohydride l-selectride , 1m in thf,lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal,lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
38721-52-7,109-99-9,1113-78-6
C12H28BLi
MFCD00011708
lithium tri-sec-butylhydroborate,l-selectridesolution,lithium tri-sec-butylborohydride solution,l-selectride™ solution,lithium tri butan-2-yl boron 1-,lithium tri-sec-butylborohydride l-selectride , 1m in thf,lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal,lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
Reduction reagent
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.8900g/mL
Color Colorless
Quantity 100mL
Linear Formula LiB[CH(CH3)C2H5]3H
Fieser 04,312; 06,348; 07,307; 08,308; 10,248; 12,286; 14,195; 15,192
Merck Index 15, 8564
Formula Weight 190.11
Physical Form Liquid
Chemical Name or Material Lithium tri-sec-butylborohydride

GHS P Statement Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May cause respiratory irritation. Reacts viole

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with

Danger

EINECSNumber : 254-101-1

TSCA : TSCA