Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tri-tert-butoxyaluminohydride, 1.1M solution in THF, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
AcroSeal™ Glass Bottle in Tin Can
17476-04-9,109-99-9
C12H28AlLiO3
MFCD00011532
17476-04-9,109-99-9
C12H28AlLiO3
MFCD00011532
Reduction reagent
Density 0.9000g/mL
Formula Weight 254.28
Physical Form Solution
Gravimetry 27 to 33% (Dry substance / 60°C)
Packaging AcroSeal™ Glass Bottle in Tin Can
Storage Information Product may develop turbidity during storage
Color Colorless
Quantity 100mL
Chemical Name or Material Lithium tri-tert-butoxyaluminohydride, 1.1M Solution in THF

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form exp

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present an

Danger