Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tri-tert-butoxyaluminohydride, 93-98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
17476-04-9
C12H28AlLiO3
254.27
MFCD00011532
17476-04-9
C12H28AlLiO3
254.27
MFCD00011532
Reduction reagent
Packaging Glass Bottle
Melting Point 319.0°C
Sublimation Point 280 °C
Infrared Spectrum Authentic
Quantity 100g
Density 0.9
Color Gray-White to White
Assay Percent Range 93-98%
Linear Formula LiAI[OC(CH3)3]3H
Fieser 01,620; 02,251; 03,188; 04,312; 10,248; 12,286; 13,342; 16,197
Solubility Information Solubility in water: reacts violently. Other solubilities: 36g/100g thf (25°C),2g/100g diethyl ether (25°C),41g/100g diglyme (25°C)
Formula Weight 254.27
Physical Form Powder
Percent Purity 93-98%
Chemical Name or Material Lithium tri-tert-butoxyaluminohydride, 93-98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger