Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tri-tert-butoxyaluminohydride, 93-98%, Thermo Scientific Chemicals

Catalog Number AC125165000
Click to view available options
Quantity:
500 g
17476-04-9
C12H28AlLiO3
254.27
MFCD00011532
17476-04-9
C12H28AlLiO3
254.27
MFCD00011532

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 319.0°C
Density 0.9
Infrared Spectrum Authentic
Assay Percent Range 93% min
Packaging Glass Bottle packed in tin can
Linear Formula LiAI[OC(CH3)3]3H
Quantity 500 g
Fieser 01,620; 02,251; 03,188; 04,312; 10,248; 12,286; 13,342; 16,197
Solubility Information Solubility in water: reacts violently. Other solubilities: 36g/100g thf (25°C),2g/100g diethyl ether (25°C),41g/100g diglyme (25°C)
Sublimation Point 280 °C
Formula Weight 254.27
Percent Purity 93 to 98%
Physical Form Powder
Chemical Name or Material Lithium tri-tert-butoxyaluminohydride, 93-98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 241-490-8

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit