Alfa Aesar™ Lithium tri-tert-butoxyaluminum hydride, 94%

Manufacturer: Alfa Aesar™ 30716-6Y

Quantity 2 x 25g

 View more versions of this product

Catalog No. AA307166Y

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 17476-04-9
    Linear Formula Li[(CH3)3CO]3AlH
    Formula Weight 254.28
    MDL Number MFCD00011532
    UN Number UN1409
    Beilstein 5796791
    Synonym Lithium hydridotris(tert-butoxy)aluminate
    Chemical Name or Material Lithium tri-tert-butoxyaluminum hydride

    Specifications

    Formula Weight 254.28
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use ... for extinction.
    Store in a dry place. Store in a closed container.
    Beilstein 5796791
    Health Hazard 2 GHS H Statement
    H228-H261-H314-H318
    Flammable solid.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    RTECS Number OJ5585000
    UN Number UN1409
    Density 1.03g/mL
    Health Hazard 1 Danger
    TSCA TSCA
    Melting Point ≈319°C (dec.)
    Physical Form Powder
    CAS 17476-04-9
    Quantity 2 x 25g
    Synonym Lithium hydridotris(tert-butoxy)aluminate
    Color White to pale gray
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METAL HYDRIDES, WATER REACTIVE, N.O.S.
    Sensitivity Moisture Sensitive
    Linear Formula Li[(CH3)3CO]3AlH
    MDL Number MFCD00011532
    Odor Odorless
    Chemical Name or Material Lithium tri-tert-butoxyaluminum hydride
    Assay 94%

    GHS H Statement
    H228-H261-H314-H318
    Flammable solid.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use ... for extinction.
    Store in a dry place. Store in a closed container.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit