Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium tri-tert-butoxyaluminum hydride, nominally 1M in THF, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 43324AD

Quantity 50mL

 View more versions of this product

Catalog No. AA43324AD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

17476-04-9
Li[(CH3)3CO]3AlH
MFCD00011532
Lithium hydridotris(tert-butoxy)aluminate
C12H28AlLiO3
254.28
UN3399
Lithium tri-tert-butoxyaluminum hydride

Specifications

Nominally 1M in THF
50mL
Colorless
ChemSeal™ bottles
Liquid
Li[(CH3)3CO]3AlH
MFCD00011532
Lithium hydridotris(tert-butoxy)aluminate
17476-04-9
0.904g/mL
−17°C (1°F)
Moisture Sensitive
C12H28AlLiO3
254.28
UN3399
Lithium tri-tert-butoxyaluminum hydride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H260-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit