Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium triethylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
800mL
Packaging:
AcroSeal™ Glass Bottle
22560-16-3,109-99-9
C6H16BLi
MFCD00011703
Lithium triethylhydroborate
22560-16-3,109-99-9
C6H16BLi
MFCD00011703
Lithium triethylhydroborate
Reduction reagent
Linear Formula LiB(C2H5)3H
Concentration 0.9 to 1.1M
Fieser 04,313; 06,348; 08,309; 09,286; 10,249; 11,304; 12,289; 17,172
Density 0.8920g/mL
Merck Index 15, 5600
Formula Weight 105.94
Physical Form Cloudy Solution
Packaging AcroSeal™ Glass Bottle
Color Colorless to Gray
Quantity 800mL
Chemical Name or Material Lithium triethylborohydride, 1M Solution in THF
Show More Show Less

GHS P Statement Highly flammable liquid and vapour. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep away from any possible contact with water, because of

Danger

EINECSNumber : 245-076-8