Alfa Aesar™ Lithium triethylborohydride, 1M in THF

As reducing agent

Manufacturer: Alfa Aesar™ 89144AE

Quantity 100mL

 View more versions of this product

Catalog No. AA89144AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22560-16-3
    Molecular Formula C6H16BLi
    Linear Formula LiB(CH2CH3)3H
    Formula Weight 105.94
    MDL Number MFCD00011703
    UN Number UN3399
    Beilstein 4151240
    Merck Index 14,5544
    Synonym Lithium triethylhydridoborate
    Chemical Name or Material Lithium triethylborohydride

    Specifications

    Formula Weight 105.94
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Molecular Formula C6H16BLi
    Density 0.892g/mL
    Health Hazard 2 GHS H Statement
    H225-H260-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    UN Number UN3399
    Sensitivity Moisture Sensitive
    MDL Number MFCD00011703
    Beilstein 4151240
    Merck Index 14,5544
    Concentration 1M in THF
    Color Colorless
    Quantity 100mL
    Physical Form Liquid
    Linear Formula LiB(CH2CH3)3H
    CAS 22560-16-3
    Synonym Lithium triethylhydridoborate
    Health Hazard 1 Danger
    Flash Point −17°C (1°F)
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    Chemical Name or Material Lithium triethylborohydride

    GHS H Statement
    H225-H260-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit