Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium triethylborohydride, 1M in THF, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 42491AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42491AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

22560-16-3
105.94
UN3399
14,5544
Lithium triethylborohydride
LiB(CH2CH3)3H
MFCD00011703
4151240
Lithium triethylhydridoborate

Specifications

22560-16-3
Liquid
105.94
MFCD00011703
4151240
Lithium triethylhydridoborate
Colorless
−17°C (1°F)
Lithium triethylborohydride
ChemSeal™ bottles
LiB(CH2CH3)3H
0.892g/mL
UN3399
14,5544
1M in THF
100mL
Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H260-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit