Alfa Aesar™ Lithium trifluoroacetate monohydrate, 97%

Manufacturer: Alfa Aesar™ 8893818

Quantity 50g

 View more versions of this product

Catalog No. AA8893818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2923-17-3
    Molecular Formula C2F3LiO2·H2O
    Linear Formula LiCO2CF3·H2O
    Formula Weight 137.97 (119.96 Anhydrous)
    MDL Number MFCD00149663
    UN Number UN2811
    Synonym Trifluoroacetic acid lithium salt
    Chemical Name or Material Lithium trifluoroacetate monohydrate

    Specifications

    Formula Weight 137.97 (119.96 Anhydrous)
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 50g
    Synonym Trifluoroacetic acid lithium salt
    Molecular Formula C2F3LiO2·H2O
    CAS 2923-17-3
    Health Hazard 2 GHS H Statement
    H300
    Fatal if swallowed.
    Color White
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00149663
    UN Number UN2811
    Sensitivity Hygroscopic
    Linear Formula LiCO2CF3·H2O
    Health Hazard 1 Danger
    Chemical Name or Material Lithium trifluoroacetate monohydrate
    Assay 97%

    GHS H Statement
    H300
    Fatal if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit