Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
79172-99-9,109-99-9
C21H46AlLiO3
380.51
LOQLYMHNMOOMHA-UHFFFAOYSA-N
lithium tris 3-ethyl-3-pentyl oxy aluminohydride
16689417
lithium;tris(3-ethylpentan-3-yloxy)aluminum(1-)
[Li+].CCC(CC)(CC)O[Al-](OC(CC)(CC)CC)OC(CC)(CC)CC
79172-99-9,109-99-9
C21H46AlLiO3
380.51
LOQLYMHNMOOMHA-UHFFFAOYSA-N
lithium tris 3-ethyl-3-pentyl oxy aluminohydride
16689417
lithium;tris(3-ethylpentan-3-yloxy)aluminum(1-)
[Li+].CCC(CC)(CC)O[Al-](OC(CC)(CC)CC)OC(CC)(CC)CC
Reduction reagent
Additional Information product may develop some turbidity
Formula Weight 380.51
Physical Form Solution
Concentration 0.40 to 0.60M
Density 0.9040g/mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.904
Color Colorless
Quantity 100mL
Flash Point −17°C
Chemical Name or Material Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
May form explosive per

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger