Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium, wire, diameter 3.2mm, 99+%, ACROS Organics™

Click to view available options
Quantity:
50g
7439-93-2
Li
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549
3028194
CHEBI:30145
lithium
[Li]
7439-93-2
Li
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
litio,litium,lithium, metallic,lithium, elemental,monohydride,unii-9fn79x2m3f,hsdb 647,hydride lih,hydrure de french,hsdb 549
3028194
CHEBI:30145
lithium
[Li]
Quantity 50g
Density 0.53g/mL
Assay Percent Range 99+%
Fieser 01,570; 03,174; 04,286; 05,376; 10,234; 11,286; 13,157; 15,184; 17,161
Merck Index 15, 5576
Formula Weight 6.94
Physical Form Wire
Percent Purity ≥99.0% (Li)
Storage Information Product turns dark gray in air
Color Silver
Chemical Name or Material Lithium
Show More Show Less