Alfa Aesar™ Lithium zirconium oxide, 94% min

Manufacturer: Alfa Aesar™ 3937736

Quantity 500g

 View more versions of this product

Catalog No. AA3937736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12031-83-3
    Formula Weight 153.1
    MDL Number MFCD00016182
    Synonym Lithium zirconate
    Chemical Name or Material Lithium zirconium oxide

    Specifications

    Formula Weight 153.1
    Mesh Size −80 Mesh
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00016182
    CAS 12031-83-3
    Synonym Lithium zirconate
    TSCA TSCA
    Color White to cream
    Quantity 500g
    Chemical Name or Material Lithium zirconium oxide
    Assay ≤94%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit