Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Litmus, Indicator (pH 5-8), for titration, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Linear Formula | CHCl3 |
Grade | Analytical Reagent |
Packaging | Glass Bottle |
Quantity | 500 g |
Chemical Name or Material | Litmus, Indicator (pH 5-8), for titration |
Safety and Handling
ShelfLife | 1620 days from date of manufacture |
EINECSNumber | 215-739-6 |
Recommended Storage | Room Temp |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction