Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Litmus, Indicator, pH 5-8, for titration, Honeywell Fluka™

Indicator, pH 5-8, for titration

Specifications

CAS Max % ≤100.0000%, ≤4.0000%
CAS Min % ≥96.0000%
Linear Formula I2
CAS 1393-92-6, 64-17-5
MDL Number MFCD00011355
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
60-048-046 View Documents Honeywell Fluka™
L7382100G
100g Glass Bottle This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
60-048-047 View Documents Honeywell Fluka™
L738225G
25g Glass Bottle This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

≤100.0000%, ≤4.0000%
I2
MFCD00011355
I2
Glass Bottle
≥96.0000%
1393-92-6, 64-17-5
Litmus
Indicator, pH 5-8, for titration
Safety and Handling

Safety and Handling

ShelfLife : 1620 days from date of manufacture

Recommended Storage : Room Temp

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit