Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Littauer Cutting Forceps, Sklar 6 in., Straight Back Blades, Straight, Smooth, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 971261
Forceps, Littauer Cutting Forceps, Sklar, 6 in., Straight Back Blades, Straight, Smooth, Stainless Steel, EA. Sklar's Littauer Cutting Forceps are a non-ratcheted, plier handle instrument used primarily for cutting nail and bone. As nail nippers they are used for nail debridement. They may also be used on delicate bones found in the facial and nasal region, or small animal (mice) veterinary procedures. The short, sharp blades are able to produce enough force to cut through thick nail and small bone, but delicate enough not to cause trauma to surrounding tissue or organs. This is particularly helpful with nail debridement which should be completed in short, small sections to avoid damage to the hyponychium. This product is straight with smooth edges and an overall length of 6 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6547