Learn More
Live Algae Cultures
$9.19
Specifications
Product Type | Green Algae Culture |
---|---|
Quantity | Each |
Sufficient For | Class of 25 students |
Catalog Number | Mfr. No. | Scientific Name | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Scientific Name | Price | Quantity & Availability | |||||
S21250
![]() |
Biological Resource Center
DCS21250 |
Protococcus |
Each for $19.68
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S21241
![]() |
Biological Resource Center
DCS S21241 |
Ulothrix |
Each for $9.19
|
|
Description
Our cultures are guaranteed to be live and healthy or we will replace them, or refund your money.
Mixed Algae Suspension Set Has five or more species in one container. Typically includes five from the following: spirogyra, mixed diatoms, Volvox, Gloeocapsa, Chlorella, and Euglena.
Representative Algae Set Includes a set of six individual cultures representing the major divisions of algae. Typically includes: Gloeocapsa, spirogyra, Chlorella, Euglena, mixed diatoms, and Volvox.
Order Info
Class size: 30 students.
Specifications
Green Algae Culture | |
Each | |
Class of 25 students |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.