Learn More
Thermo Scientific™ Live Cell Reagents
Monitor cell viability and cell morphology in live cells.
Products 3
Catalog Number Mfr. No. Description Quantity Target Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Target Price Quantity & Availability  
84-101-01
View Documents
Thermo Scientific™
8410101
Live Cell Blue 5 x 100µg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
84-100-01
View Documents
Thermo Scientific™
8410001
Live Cell Green 5 x 100µg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
84-102-01
View Documents
Thermo Scientific™
8410201
Live Cell Violet 5 x 100µg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Monitor cell viability and cell morphology in live cells with Thermo Scientific™ Live Cell Stains.
The hydrolysis of the non-fluorescent Cellomics™ Live Cell Stain Blue by intracellular esterases generates the strongly fluorescent hydrophilic Cellomics Live Cell Stain Blue that is well retained in the cell cytoplasm. The esterase activity is proportional to the number of viable cells, and the fluorescence intensity generated from the esterase-catalyzed hydrolysis of the stains can also be used to measure the cell number.
Highlights:
  • Non-fluorescent dye becomes strongly fluorescent upon entering live cells
  • Hydrophobic compound easily permeates intact live cells
  • Blue fluorescence is ideal for multiplexing with other fluorescence staining of cells
  • Readily adapted for high throughput assays in a wide variety of fluorescence platforms such as microplate assays, immunocytochemistry and flow cytometry
  • Useful for a variety of studies, including cell adhesion, chemotaxis, multidrug resistance, cell viability, apoptosis and cytotoxicity
  • Suitable for proliferating and non-proliferating cells, and can be used for both suspension and adherent cells