Learn More
BioVision IncSupplier Diversity Partner Live-Dead Cell Staining Kit: Live-Dead Cell Staining Kit: Distinguishing between live and dead cells within 20 min. Ready-to-Use Reagents. Detection Method: Fluorescent Microscopy. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K501100
The assay range is 0.156–10 ng/ml ANGPTL3/ml. The lowest level of ANGPTL3 that can be detected by this assay is 75 pg/ml. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9758