Learn More
Aviva Systems Biology Corp LIX1L Antibody - N-terminal region (ARP53201_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIX1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LIX1L. Alternate names: DKFZp762F237, MGC46719ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60968