Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIX1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548405

Catalog No. PA548405

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rat based on sequence homology.

Specifications

Specifications

LIX1L
Polyclonal
PBS with 0.09% sodium azide
Q8IVB5, Q8BQ89
LIX1-like protein, LIX1L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
128077, 280411
Western Blot
Unconjugated
LIX1L
Liquid
LIX1L
KLH conjugated synthetic peptide between 160-189 amino acids from the Central region of human LIX1L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit