Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ LKB1/STK11 Human Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$344.00 - $976.00

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-Page
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% Glycerol
Gene ID (Entrez) 6794
Molecular Weight TMW: 51kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25195905
View Documents
Novus Biologicals™
NBP2519590.05MG
0.05mg
Each for $344.00
Add to cart
 
NBP25195925
View Documents
Novus Biologicals™
NBP2519590.25MG
0.25mg
Each for $976.00
Add to cart
 
Description

Description

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-433 of Human LKB1/STK11 The Recombinant Human LKB1/STK11 Protein is derived from E. coli. The Recombinant Human LKB1/STK11 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.5mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 10% Glycerol
TMW: 51kDa
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
LKB1/STK11
Yes
SDS-Page
6794
>85%, by SDS-PAGE
EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11
Unlabeled
SDS
Product Certifications

For research use only and is not approved for use in humans or in clinical diagnosis

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit