Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LKB1/STK11 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI0A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271300JF549

Catalog No. N271300J549


Add to cart

Description

Description

LKB1/STK11 Monoclonal antibody specifically detects LKB1/STK11 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

LKB1/STK11
OTI0A3
Immunohistochemistry
Affinity Purified, Conjugated
STK11
Recombinant protein fragment corresponding to amino acids 1-236 of human LKB1/STK11 (NP_000446) produced in E.coli.
Immunogen affinity purified
Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase
Primary
6794.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit