Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LLGL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550698
Description
The antibody detects endogenous levels of total LLGL1 protein.
LLGL1 Polyclonal antibody specifically detects LLGL1 in Human samples. It is validated for Western Blot.Specifications
LLGL1 | |
Polyclonal | |
Unconjugated | |
LLGL1 | |
AI325176; DLG4; HUGL; Hugl protein; HUGL1; Hugl-1; Human homolog to the D-lgl gene protein; lethal giant larvae homolog 1; lethal giant larvae homolog 1 (Drosophila); lethal giant larvae homolog 1, scribble cell polarity complex component; lethal(2) giant larvae protein homolog 1; Lgl1; LLGL; Llgl1; LLGL1, scribble cell polarity complex component; Llglh; Mgl1; Mgl-1; Mlgl; Rgl1; Rgl-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3996 | |
-20°C | |
Liquid |
Western Blot | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q15334 | |
LLGL1 | |
Synthetic peptide corresponding to residues near the C terminal of human lethal giant larvae homolog 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction