Learn More
Selleck Chemical LLC LLY-283 5mg
SDP
Supplier:  Selleck Chemical LLC S88835MG
Encompass_Preferred
LLY-283 is a novel and selective inhibitor of protein arginine methyltransferase 5 (PRMT5). LLY-283 inhibits PRMT5 enzymatic activity in vitro and in cells with IC50 of 22 nM and 25 nM, respectively. LLY-283 shows antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3417
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.