Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LM 22A4, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
37988-18-4
C15H21N3O6
339.348
RGWJKANXFYJKHN-UHFFFAOYSA-N
lm 22a4, potent trkb agonist,n,n',ntris 2-hydroxyethyl-1,3,5-benzenetricarboxamide,1-n,3-n,5-n-tris 2-hydroxyethyl benzene-1,3,5-tricarboxamide,n1,n3,n5-tris 2-hydroxyethyl benzene-1,3,5-tricarboxamide
2054170
1-N,3-N,5-N-tris(2-hydroxyethyl)benzene-1,3,5-tricarboxamide
C1=C(C=C(C=C1C(=O)NCCO)C(=O)NCCO)C(=O)NCCO
This item is not returnable. View return policy
37988-18-4
C15H21N3O6
339.348
RGWJKANXFYJKHN-UHFFFAOYSA-N
lm 22a4, potent trkb agonist,n,n',ntris 2-hydroxyethyl-1,3,5-benzenetricarboxamide,1-n,3-n,5-n-tris 2-hydroxyethyl benzene-1,3,5-tricarboxamide,n1,n3,n5-tris 2-hydroxyethyl benzene-1,3,5-tricarboxamide
2054170
1-N,3-N,5-N-tris(2-hydroxyethyl)benzene-1,3,5-tricarboxamide
C1=C(C=C(C=C1C(=O)NCCO)C(=O)NCCO)C(=O)NCCO

Potent TrkB agonist

Potent tropomyosin-related kinase B (TrkB) agonist (IC50 = 47 nM). Induces activation of Trk, Akt and ERK in mouse hippocampus and striatum. Reverses deficits in motor task learning in mice following traumatic brain injury; restores respiratory function in a rat model of Rett syndrome.
Quantity 50mg
Formula Weight 339.34
Percent Purity >98%
Chemical Name or Material LM 22A4

Recommended Storage : Store at +4°C