Learn More
LM 22A4, Tocris Bioscience™
Potent TrkB agonist
Manufacturer:  Tocris Bioscience 4607/25
 View more versions of this product
Catalog No. 46-072-5
Add to cart
This item is not returnable. View return policy
Description
Description
Potent tropomyosin-related kinase B (TrkB) agonist. Induces activation of Trk, Akt and ERK in mouse hippocampus and striatum and exhibits neurotrophic activity. Reverses deficits in motor task learning in mice following traumatic brain injury; restores respiratory function in a rat model of Rett syndrome.
Specifications
Specifications
N,N',N''Tris(2-hydroxyethyl)-1,3,5-benzenetricarboxamide
>98%
Observed MW: 339.34
37988-18-4
C15H21N3O6
25mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C