Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMAN1 Mouse anti-Human, Rat, Canine, Monkey, Alexa Fluor™ 405, Clone: OTI1A8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271152AF405

Catalog No. N271152A405


Add to cart

Description

Description

LMAN1 Monoclonal antibody specifically detects LMAN1 in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

LMAN1
OTI1A8
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
LMAN1
Full length recombinant protein of human LMAN1 (NP_005561) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
ERGIC-53, ERGIC53ER-Golgi intermediate compartment 53 kDa protein, F5F8Dendoplasmic reticulum-golgi intermediate compartment protein 53, FMFD1, Gp58, intracellular mannose specific lectin, Intracellular mannose-specific lectin MR60, Lectin mannose-binding 1, lectin, mannose-binding, 1, MCFD1coagulation factor V-factor VIII combined deficiency, MR60, protein ERGIC-53
Mouse
IgG1
0.1 mL
Primary
3998.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit