Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMAN1 Mouse anti-Canine, Human, Clone: OTI1E3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525346

Catalog No. PIMA525346


Add to cart

Description

Description

The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
Specifications

Specifications

LMAN1
Monoclonal
0.24 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P49257
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58, ER-Golgi intermediate compartment 53 kDa protein, endoplasmic reticulum-golgi intermediate compartment protein 53, intracellular mannose specific lectin, intracellular mannose-specific lectin MR60, protein ERGIC-53
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3998, 476186
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1E3
Unconjugated
LMAN1
Liquid
LMAN1
Full length human recombit protein of LMAN1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit