Learn More
Laxco™ LMC-4000 Series Clinical Microscope, Microbiology Fluorescence Configuration
The Laxco™ LMC-4000 Series Fluorescence Upright Microscope System combines research-grade components and ergonomic features designed for the demands of the clinical environment. Available class leading features include LED fluorescence illuminator, tilting ergonomic head, infinity corrected plan achromat or plan fluor objectives, 5-position or 6-position nosepiece and LED illumination.
Manufacturer:  Laxco™ LMC4MC4
 View more versions of this product
Catalog No. LMC4MC4
Add to cart
Description
Description
Key Features
  • Y-Shaped body for maximum stability
  • Carrying handle
  • Stable main body
  • Brightfield, Darkfield, Polarization, Metallurgical, Phase Contrast, and Fluorescence contrast methods
  • High contrast, broad beam optical design
  • Modular design allows for flexibility in system configuration - 100% customizable for every need
LED Fluorescence
  • 25,000 hour life span
  • No alignment needed
  • LED wave length specific to filter set
  • Adjustable light intensity
  • High signal-to-noise ratio
  • Stable light excitation energy
  • No warm-up or cool down time
  • Produces no harmful UV light
  • Powered by optional battery pack
Optical System
  • Infinity corrected optical system
  • Plan Achromat objectives standard, Plan Fluor (optional)
  • Wide range of magnifications: 2X, 4X, 10X, 40X, 60X, 100X
  • Anti-reflective coatings on optics
  • Sharp images with bright, natural color
  • Parcentric and parfocal objectives
Filter Sets
  • LMC4MC3: [1] DAPI/Hoechst/AMCA/AMCA-X Blue/Alexa Fluor 350 (EX: 365/25, EM:460/25), [2] EGFP/GFP/FITC (EX: 470/20, EM: 525/20), [3] Texas RED/XCy3.5/Mito, Tracker Red (EX: 470/20, EM: 525/20
  • LMC4MC4: [1] DAPI/Hoechst/AMCA/AMCA-X Blue/Alexa Fluor 350 (EX: 365/25, EM:460/25), [2] EGFP/GFP/FITC (EX: 470/20, EM: 525/20), [3] Texas RED/XCy3.5/Mito, Tracker Red (EX: 470/20, EM: 525/20
Specifications
Specifications
Tilting Trinocular
Brightfield, LED Fluorescence
4X, 10X, 40X, 100X Oil Plan Fluor
3w LED
1.2 N.A. Condenser with Swing-out Lens
Infinity, 22mm FOV