Learn More
Laxco™ LMC-4000 Series Clinical Microscope, Pathology/Histology Configuration
The Laxco™ LMC-4000 Series Upright Microscope System combines research-grade components and ergonomic features designed for the demands of the clinical environment. Available class leading features include tilting ergonomic head, infinity corrected plan achromat or plan fluor objectives, 5-position or 6-position nosepiece and LED illumination.
$5858.04 - $17522.60
Specifications
Eyepiece Plan 10X/20mm
Optical Design Infinity, 22mm FOV
Condenser Type 0.9 N.A. Condenser with Swing-out Lens
Lighting Type 5w LED
Product Type 6 POS
View More Specs
Products 8
Catalog Number Mfr. No. Head Style Objective Types Model Price Quantity & Availability  
Catalog Number Mfr. No. Head Style Objective Types Model Price Quantity & Availability  
LMC4PC1
View Documents
Laxco™
LMC4PC1
Trinocular 1.25X APO, 4X, 10X, 20X, 40X Plan Fluor, 60X Plan ACH Brightfield Each for $13,103.59
Add to cart
LMC4PC2
View Documents
Laxco™
LMC4PC2
Tilting Trinocular 1.25X APO, 4X, 10X, 20X, 40X Plan Fluor, 60X Plan ACH Brightfield Each for $16,710.83
Add to cart
LMC4PC3
View Documents
Laxco™
LMC4PC3
Trinocular 2X, 4X, 10X, 20X, 40X, 60X Plan Achromat Brightfield Each for $5,858.04
Add to cart
LMC4PC4
View Documents
Laxco™
LMC4PC4
Tilting Trinocular 2X, 4X, 10X, 20X, 40X, 60X Plan Achromat Brightfield Each for $9,465.27
Add to cart
LMC4PC11
View Documents
Laxco™
LMC4PC11
Trinocular 1.25X APO, 4X, 10X, 20X, 40X Plan Fluor, 60X Plan ACH Brightfield with Gout Kit Each for $13,915.36
Add to cart
LMC4PC12
View Documents
Laxco™
LMC4PC12
Tilting Trinocular 1.25X APO, 4X, 10X, 20X, 40X Plan Fluor, 60X Plan ACH Brightfield with Gout Kit Each for $17,522.60
Add to cart
LMC4PC13
View Documents
Laxco™
LMC4PC13
Trinocular 2X, 4X, 10X, 20X, 40X, 60X Plan Achromat Brightfield with Gout Kit Each for $6,669.81
Add to cart
LMC4PC14
View Documents
Laxco™
LMC4PC14
Tilting Trinocular 2X, 4X, 10X, 20X, 40X, 60X Plan Achromat Brightfield with Gout Kit Each for $10,277.07
Add to cart
Description
Description
Key Features
  • Large, sturdy base designed for the lab environment
  • Rack and pinion focusing mechanism for smooth focus control
  • Rackless, X/Y mechanical stage with adjustable tension
  • Reversed quintuple nosepiece
  • High contrast, borad beam optical desing
  • Full Koehler illumination with field and iris diaphragm
  • Modular design allows for flexibility in system configuration - 100% customizable for every application
  • LED pointer
  • 12V/100w illumination
  • Supports up to 5 heads
Optical System
  • Infinity corrected optical system
  • Plan Achromat objectives stand, Plan Fluor optional
  • Wide range of magnifications: 2X, 4X, 10X, 20X, 40X, 50X Oil, 60X, and 100X
  • Anti-reflective coatings on optics
  • Sharp images with color corrected optics
  • Parcentric and parfocal objectives
Specifications
Specifications
Plan 10X/20mm
0.9 N.A. Condenser with Swing-out Lens
6 POS
Infinity, 22mm FOV
5w LED