Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LMCD1 Monoclonal Antibody (OTI1H5)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525603

Catalog No. PIMA525603


Only null left
Add to Cart

Description

Description

LMCD1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

LMCD1 encodes a member of the LIM-domain family of zinc finger proteins. The encoded protein contains an N-terminal cysteine-rich domain and two C-terminal LIM domains. The presence of LIM domains suggests involvement in protein-protein interactions. The protein may act as a co-regulator of transcription along with other transcription factors. Alternate splicing results in multiple transcript variants of this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LMCD1
Monoclonal
0.8 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NZU5
LMCD1
Full length human recombinant protein of LMCD1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI1H5
Unconjugated
LMCD1
AW455500; AW553074; dyxin; LIM and cysteine rich domains 1; LIM and cysteine-rich domains 1; LIM and cysteine-rich domains protein 1; LIMD1; LMCD1
Mouse
Affinity chromatography
RUO
29995
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.