Learn More
Aviva Systems Biology Corp LMO2 Antibody - N-terminal region (P100952_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100952_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LMO2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LMO2. Alternate names: RBTN2, RBTNL1, RHOM2, TTG2ApplicationWB. Species reactivity: Dog, Guinea Pig, Mouse, Rabbit, Rat
Catalog No. 50-014-62796