Learn More
LMO2 Mouse anti-Human, DyLight 755, Clone: 4D8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237442IR
Catalog No. NBP237442IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
LMO2
4D8
Western Blot, Flow Cytometry, ELISA
Purified
LMO2
Purified recombinant fragment of human LMO2 (AA: 1-158) expressed in E. Coli.
Protein A or G purified
Angiogenesis, Cancer
Primary
4005
ELISA, Flow Cytometry, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human