Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO2 Rabbit anti-Human, Clone: LMO2/3147R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen LMO2
Clone LMO2/3147R
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121875
View Documents
Novus Biologicals
NBP30720110UG
100 μg
Each for $406.52
Add to cart
 
NB121876
View Documents
Novus Biologicals
NBP30720120UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications

Specifications

LMO2
0.2 mg/ml
Flow Cytometry, Immunofluorescence
Purified
RUO
Human
Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2
Recombinant fragment (around aa 23-140) human LMO2 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
LMO2/3147R
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Rabbit
Angiogenesis, Cancer
10 mM PBS with 0.05% BSA
4005
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit