Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMO7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen LMO7
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB124858
View Documents
Novus Biologicals
NBP30997825UL
25 μg
Each for $106.19
Add to cart
 
NB124857
View Documents
Novus Biologicals
NBP309978100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

LMO7 Polyclonal antibody specifically detects LMO7 in Human samples. It is validated for Western Blot
Specifications

Specifications

LMO7
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
4008
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
FBX20F-box only protein 20, KIAA0858FBXO20F-box protein Fbx20, LIM domain 7, LIM domain only 7, LIM domain only protein 7, LMO-7, LOMP, zinc-finger domain-containing protein
The immunogen is a synthetic peptide directed towards the middle region of human LMO7 (NP_001293009.1). Peptide sequence VEPKTALPFNRFLPNKSRQPSYVPAPLRKKKPDKHEDNRRSWASPVYTEA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit